2-Nitrophenyl octyl ether ≥98.0%
Supplier: TCI America
Synonyms:
1-Nitro-2-(n-octyloxy)benzene
, 2-(n-Octyloxy)nitrobenzene
, 1-Nitro-2-(octyloxy)benzene
S0380-1G
S0380-10G
TCS0380-1GEA
32.48
USD
TCS0380-1G
TCS0380-10G
2-Nitrophenyl octyl ether ≥98.0%
2-Nitrophenyl octyl ether
[Matrix for FABMS and liquid SIMS]
CAS Number: 37682-29-4
MDL Number: MFCD00014693
Molecular Formula: C14H21NO3
Molecular Weight: 251.33
Purity/Analysis Method:
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 198
Flash Point (°C): 113
Specific Gravity (20/20): 1.04
Formula:
C₁₄H₂₁NO₃ MW: 251.32 g/mol Boiling Pt: 198 °C Density: 1.04 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00014693 CAS Number: 37682-29-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...