8-Quinolinecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Quinoline-8-carboxylic acid
Q0066-1G
TCQ0066-1GEA
104.55
USD
TCQ0066-1G
8-Quinolinecarboxylic acid ≥98.0% (by GC, titration analysis)
8-Quinolinecarboxylic acid
CAS Number: 86-59-9
MDL Number: MFCD00047619
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method:
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 189
Formula:
C₁₀H₇NO₂ MW: 173.17 g/mol Melting Pt: 189 °C |
MDL Number:
MFCD00047619 CAS Number: 86-59-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...