2-Pentylthiophene ≥98.0%

Supplier: TCI America

P1835-5G
TCP1835-5GEA 42 USD
TCP1835-5G
2-Pentylthiophene ≥98.0%
2-Pentylthiophene

CAS Number: 4861-58-9
MDL Number: MFCD00041017
Molecular Formula: C9H14S
Molecular Weight: 154.27
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 204
Flash Point (°C): 75
Specific Gravity (20/20): 0.95


Product Categories: Materials Chemistry, Building Blocks for Semiconducting Polymers
Other Product Names / Synonyms: 2-Amylthiophene

Formula: C₉H₁₄S
MW: 154.28 g/mol
Boiling Pt: 204 °C
Density: 0.95 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00041017
CAS Number: 4861-58-9
https://us.vwr.com/stibo/web/std.lang.all/31/27/28993127.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR