Phthalocyanine ≥98.0% (by total nitrogen basis), purified by sublimation

Supplier: TCI America

Synonyms: 29H,31H-PhthalocyaninePigment Blue 16

P1795-1G
TCP1795-1GEA 149 USD
TCP1795-1G
Phthalocyanine ≥98.0% (by total nitrogen basis), purified by sublimation
Phthalocyanine

CAS Number: 574-93-6
MDL Number: MFCD00005085
Molecular Formula: C32H18N8
Molecular Weight: 514.55
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Color: Deep Blue


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Organic Transistor Materials, Molecular Conductors, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Highly Purified Reagents, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Pigment Blue 16

Formula: C₃₂H₁₈N₈
MDL Number: MFCD00005085
CAS Number: 574-93-6
https://us.vwr.com/stibo/web/std.lang.all/30/97/28993097.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR