5,7,12,14-Pentacenetetrone ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,7,12,14-Tetraoxo-5,7,12,14-tetrahydropentacene

P1710-25G P1710-5G
TCP1710-25GEA 311 USD
TCP1710-25G TCP1710-5G
5,7,12,14-Pentacenetetrone ≥95.0% (by HPLC)
5,7,12,14-Pentacenetetrone

CAS Number: 23912-79-0
MDL Number: MFCD00075209
Molecular Formula: C22H10O4
Molecular Weight: 338.32
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 400


Other Product Names / Synonyms: 5,7,12,14-Tetraoxo-5,7,12,14-tetrahydropentacene

Formula: C₂₂H₁₀O₄
Melting Pt: 400 °C
MDL Number: MFCD00075209
CAS Number: 23912-79-0
https://us.vwr.com/stibo/web/std.lang.all/30/40/28993040.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR