1-Propylpiperazine dihydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1645-25G
TCP1645-25GEA 372.4 USD
TCP1645-25G
1-Propylpiperazine dihydrobromide ≥98.0% (by titrimetric analysis)
1-Propylpiperazine dihydrobromide

CAS Number: 64262-23-3
Molecular Formula: C7H16N2
Molecular Weight: 290.04
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 254

Formula: C₇H₁₆N₂
Melting Pt: 254 °C
MDL Number: MFCD00051992
CAS Number: 64262-23-3
https://us.vwr.com/stibo/web/std.lang.all/21/89/28992189.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR