(Phenylthio)acetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 2-(Phenylthio)acetonitrile

P1602-5G
TCP1602-5GEA 53 USD
TCP1602-5G
(Phenylthio)acetonitrile ≥98.0%
(Phenylthio)acetonitrile

CAS Number: 5219-61-4
MDL Number: MFCD00019831
Molecular Formula: C8H7NS
Molecular Weight: 149.21
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 146
Specific Gravity (20/20): 1.14

Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 146 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00019831
CAS Number: 5219-61-4
https://us.vwr.com/stibo/web/std.lang.all/21/57/28992157.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR