Polystyrene resin, cross-linked with 1% DVB (200-400mesh)

Supplier: TCI America

P1379-25G
TCP1379-025GEA 53 USD
TCP1379-025G
Polystyrene resin, cross-linked with 1% DVB (200-400mesh)
Polystyrene resin

MDL Number: MFCD00084450


Product Categories: Synthetic Organic Chemistry, Resins (Combinatorial Chemistry)

MDL Number: MFCD00084450
CAS Number: 9003-70-7
https://us.vwr.com/stibo/web/std.lang.all/20/29/28992029.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR