Methyl pentafluoropropionate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2,2,3,3,3-pentafluoropropanoate

P1352-25G P1352-5G
TCP1352-5GEA 27 USD
TCP1352-5G TCP1352-025G
Methyl pentafluoropropionate ≥98.0%
Methyl pentafluoropropionate

CAS Number: 378-75-6
MDL Number: MFCD00000430
Molecular Formula: C4H3F5O2
Molecular Weight: 178.06
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Flash Point (°C): 6
Specific Gravity (20/20): 1.40


Other Product Names / Synonyms: Methyl Perfluoropropionate, Pentafluoropropionic Acid Methyl Ester, Perfluoropropionic Acid Methyl Ester

Formula: C₄H₃F₅O₂
MW: 178.06 g/mol
Boiling Pt: 60 °C
Density: 1.40 (20 °C)
Flash Pt: 6 °C
UN: 3272
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/20/00/28992000.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR