Perfluoro(allylbenzene) ≥98.0%

Supplier: TCI America

P1343-5G
TCP1343-005GEA 44 USD
TCP1343-005G
Perfluoro(allylbenzene) ≥98.0%
Perfluoro(allylbenzene)

CAS Number: 67899-41-6
MDL Number: MFCD00153225
Molecular Formula: C9F10
Molecular Weight: 298.08
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.71

Formula: C₉F₁₀
Density: 1.71 (20 °C)
MDL Number: MFCD00153225
CAS Number: 67899-41-6
https://us.vwr.com/stibo/web/std.lang.all/19/94/28991994.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR