2-Phenyl-4-penten-2-ol ≥97.0%

Supplier: TCI America

P1330-5G
TCP1330-005GEA 89 USD
TCP1330-005G
2-Phenyl-4-penten-2-ol ≥97.0%
2-Phenyl-4-penten-2-ol

CAS Number: 4743-74-2
MDL Number: MFCD00060886
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 100
Flash Point (°C): 66
Specific Gravity (20/20): 0.99

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Boiling Pt: 100 °C
Density: 0,99 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00060886
CAS Number: 4743-74-2

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR