4-Phenoxybenzophenone

Supplier: TCI America

P1328-5G
TCP1328-005GEA 131 USD
TCP1328-005G
4-Phenoxybenzophenone
4-Phenoxybenzophenone

CAS Number: 6317-73-3
MDL Number: MFCD00017553
Molecular Formula: C19H14O2
Molecular Weight: 274.32
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 64

Formula: C₁₉H₁₄O₂
Melting Pt: 64 °C
MDL Number: MFCD00017553
CAS Number: 6317-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR