2-Phenylquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Phenyl-4-quinolinecarboxylic acid
P1301-25G
TCP1301-025GEA
100.49
USD
TCP1301-025G
2-Phenylquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2-Phenylquinoline-4-carboxylic acid
CAS Number: 132-60-5
MDL Number: MFCD00006750
Molecular Formula: C16H11NO2
Molecular Weight: 249.27
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 215
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: Cinchophen, 2-Phenylcinchoninic Acid
Formula:
C₁₆H₁₁NO₂ MW: 249.27 g/mol Melting Pt: 215 °C |
MDL Number:
MFCD00006750 CAS Number: 132-60-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...