α-Phenylcinnamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1300-25G
TCP1300-025GEA 308.56 USD
TCP1300-025G
α-Phenylcinnamic acid ≥98.0% (by titrimetric analysis)
α-Phenylcinnamic acid

CAS Number: 91-48-5
MDL Number: MFCD00004252
Molecular Formula: C15H12O2
Molecular Weight: 224.26
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 175


Other Product Names / Synonyms: 2,3-Diphenylacrylic Acid

Formula: C₁₅H₁₂O₂
Melting Pt: 175 °C
MDL Number: MFCD00004252
CAS Number: 91-48-5
https://us.vwr.com/stibo/web/std.lang.all/19/46/28991946.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR