4-Phenoxybutyric acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
P1285-25G
TCP1285-025GEA
94
USD
TCP1285-025G
4-Phenoxybutyric acid ≥97.0% (by titrimetric analysis)
4-Phenoxybutyric acid
CAS Number: 6303-58-8
MDL Number: MFCD00042656
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method:
Form: Crystal
Color: Slightly Pale Yellow
Boiling point (°C): 170
Melting point (°C): 64
Formula:
C₁₀H₁₂O₃ Boiling Pt: 170 °C Melting Pt: 64 °C |
MDL Number:
MFCD00042656 CAS Number: 6303-58-8 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...