(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-Phe-OMe·HCl
P1278-25G
TCP1278-025GEA
35
USD
TCP1278-025G
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride ≥98.0% (by titrimetric analysis)
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
CAS Number: 7524-50-7
MDL Number: MFCD00012489
Molecular Formula: C10H13NO2
Molecular Weight: 215.68
Purity/Analysis Method:
Form: Crystal
Color: White
Specific rotation [a]20/D: 38 deg (C=2, EtOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Phe-OMe.HCl
Formula:
C₁₀H₁₄ClNO₂ MW: 215.68 g/mol |
MDL Number:
MFCD00012489 CAS Number: 7524-50-7 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...