Phthalimidoacetaldehyde diethyl acetal ≥95.0%
Supplier: TCI America
Synonyms:
2-(2,2-Diethoxyethyl)isoindoline-1,3-dione
, Phthalimidoacetaldehyd Diethylacetal
P1229-25G
TCP1229-025GEA
114.45
USD
TCP1229-025G
Phthalimidoacetaldehyde diethyl acetal ≥95.0%
Phthalimidoacetaldehyde diethyl acetal
CAS Number: 78902-09-7
MDL Number: MFCD00005901
Molecular Formula: C14H17NO4
Molecular Weight: 263.29
Purity/Analysis Method:
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 72
Other Product Names / Synonyms: N-(2,2-Diethoxyethyl)phthalimide
Formula:
C₁₄H₁₇NO₄ MW: 263.30 g/mol Melting Pt: 72 °C |
MDL Number:
MFCD00005901 CAS Number: 78902-09-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...