Ethyl pentafluoropropionate ≥98.0%

Supplier: TCI America

P1123-25G P1123-5G
TCP1123-025GEA 93.18 USD
TCP1123-025G TCP1123-5G
Ethyl pentafluoropropionate ≥98.0%
Ethyl pentafluoropropionate

CAS Number: 426-65-3
MDL Number: MFCD00000431
Molecular Formula: C5H5F5O2
Molecular Weight: 192.09
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 76
Flash Point (°C): 2
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: Pentafluoropropionic Acid Ethyl Ester

Formula: C₅H₅F₅O₂
MW: 192.09
Boiling Pt: 76 °C
Density: 1.30 (20 °C)
Flash Pt: 2 °C
UN: 3272
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/17/52/28991752.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR