1-(2-Pyrimidyl)piperazine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(1-Piperazinyl)pyrimidine Dihydrochloride

P1119-25G
TCP1119-025GEA 174 USD
TCP1119-025G
1-(2-Pyrimidyl)piperazine dihydrochloride ≥98.0% (by HPLC, titration analysis)
1-(2-Pyrimidyl)piperazine dihydrochloride

CAS Number: 94021-22-4
MDL Number: MFCD00012779
Molecular Formula: C8H12N4
Molecular Weight: 237.13
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Flash Point (°C): 240


Other Product Names / Synonyms: 2-(1-Piperazinyl)pyrimidine Dihydrochloride

Formula: C₈H₁₂N₄
Flash Pt: 240 °C
MDL Number: MFCD00012779
CAS Number: 94021-22-4
https://us.vwr.com/stibo/web/std.lang.all/17/48/28991748.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR