Piperonylacetone ≥98.0%

Supplier: TCI America

Synonyms: 4-(3,4-Methylenedioxy)phenyl-2-butanone

P1089-25G
TCP1089-025GEA 119.9 USD
TCP1089-025G
Piperonylacetone ≥98.0%
Piperonylacetone

CAS Number: 55418-52-5
MDL Number: MFCD00016910
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 165
Melting point (°C): 51


Other Product Names / Synonyms: 4-(1,3-Benzodioxol-5-yl)-2-butanone, 4-(3,4-Methylenedioxyphenyl)-2-butanone

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR