3,4-(Methylenedioxy)benzylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Benzo[d][1,3]dioxol-5-ylmethanaminePiperonylamine

P1057-25G
TCP1057-025GEA 195.33 USD
TCP1057-025G
3,4-(Methylenedioxy)benzylamine ≥97.0% (by GC, titration analysis)
3,4-(Methylenedioxy)benzylamine

CAS Number: 2620-50-0
MDL Number: MFCD00005840
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 132
Flash Point (°C): 110
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: 5-(Aminomethyl)-1,3-benzodioxole, (1,3-Benzodioxol-5-yl)methylamine, 3,4-Methylenedioxybenzylamine

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Boiling Pt: 132 °C
Density: 1.22 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005840
CAS Number: 2620-50-0
https://us.vwr.com/stibo/web/std.lang.all/16/92/28991692.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR