Dilithium phthalocyanine ≥93.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Phthalocyanine Dilithium

P1049-1G
TCP1049-001GEA 96.22 USD
TCP1049-001G
Dilithium phthalocyanine ≥93.0% (by total nitrogen basis)
Dilithium phthalocyanine

CAS Number: 25510-41-2
MDL Number: MFCD00059094
Molecular Formula: C32H16Li2N8
Molecular Weight: 526.42
Purity/Analysis Method: 93.0% (N)
Form: Crystal
Color: Bluish Gray
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Molecular Conductors, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Phthalocyanine Dilithium

Formula: C₃₂H₁₆Li₂N₈
MW: 526.42 g/mol
MDL Number: MFCD00059094
CAS Number: 25510-41-2
https://us.vwr.com/stibo/web/std.lang.all/16/83/28991683.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR