Magnesium(II) phthalocyanine
Supplier: TCI America
Synonyms:
Phthalocyanine Magnesium(II)
P1018-1G
TCP1018-001GEA
126
USD
TCP1018-001G
Magnesium(II) phthalocyanine
Magnesium(II) phthalocyanine
CAS Number: 1661-03-6
MDL Number: MFCD00041953
Molecular Formula: C32H16MgN8
Molecular Weight: 536.84
Form: Crystal
Color: Deep Blue
Other Product Names / Synonyms: Phthalocyanine Magnesium(II)
Formula:
C₃₂H₁₆MgN₈ MW: 536.84 g/mol |
MDL Number:
MFCD00041953 CAS Number: 1661-03-6 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...