4-Pentylcyclohexanone ≥95.0%
Supplier: TCI America
P0883-5G
TCP0883-5GEA
51
USD
TCP0883-5G
4-Pentylcyclohexanone ≥95.0%
4-Pentylcyclohexanone
CAS Number: 61203-83-6
MDL Number: MFCD00059581
Molecular Formula: C11H20O
Molecular Weight: 168.28
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: 4-Amylcyclohexanone
Formula:
C₁₁H₂₀O MW: 168.28 g/mol Density: 0.89 (20 °C) |
UN:
3082 ADR: 9,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...