4-Piperidone monohydrate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Piperidin-4-one hydrate hydrochloride4-Azacyclohexanone Monohydrate Hydrochloride4,4-Dihydroxypiperidine Hydrochloride4-Piperidinone monohydrate hydrochloride4,4-Piperidinediol Hydrochloride4-Ketopiperidine Monohydrate Hydrochloride

P0842-25G P0842-100G P0842-500G
TCP0842-025GEA 72.73 USD
TCP0842-025G TCP0842-100G TCP0842-500G
4-Piperidone monohydrate hydrochloride ≥98.0% (by titrimetric analysis)
4-Piperidone monohydrate hydrochloride

CAS Number: 40064-34-4
MDL Number: MFCD00012776
Molecular Formula: C5H9NO
Molecular Weight: 153.61
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: 4-Azacyclohexanone Monohydrate Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4,4-Piperidinediol Hydrochloride

Formula: C₅H₉NO
Melting Pt: 100 °C
MDL Number: MFCD00012776
CAS Number: 40064-34-4
https://us.vwr.com/stibo/web/std.lang.all/15/10/28991510.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR