O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: PFBHA.HCl

P0822-1G P0822-5G
TCP0822-001GEA 136.68 USD
TCP0822-001G TCP0822-005G
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥98.0% (by HPLC, total nitrogen)
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride

[for Oxime Preparation]
CAS Number: 57981-02-9
MDL Number: MFCD00012953
Molecular Formula: C7H4F5NO
Molecular Weight: 249.57
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 215


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Analytical Science, Others (GC Derivatizing Reagents)
Other Product Names / Synonyms: PFBHA.HCl

Formula: C₇H₄F₅NO
Melting Pt: 215 °C
MDL Number: MFCD00012953
CAS Number: 57981-02-9
https://us.vwr.com/stibo/web/std.lang.all/14/92/28991492.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR