Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

TCIAP0696

P0696-25G P0696-500G
TCP0696-025GEA 21.09 USD InStock
TCP0696-025G TCP0696-500G
TCIAP0696
CAS Number: 9003-39-8
Molecular Formula: C6H9NO
Molecular Weight:
Form: Liquid
Color: Colorless

Product Categories: Materials Chemistry, Polymer Research Reagents

Order Now
 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty