2-Phenoxypropionyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P0638-25G P0638-500G
TCP0638-25GEA 70.02 USD
TCP0638-25G TCP0638-500G
2-Phenoxypropionyl chloride ≥98.0% (by GC, titration analysis)
2-Phenoxypropionyl chloride

CAS Number: 122-35-0
MDL Number: MFCD00018810
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 100
Specific Gravity (20/20): 1.18

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 106 °C
Density: 1.18 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00018810
CAS Number: 122-35-0
https://us.vwr.com/stibo/web/std.lang.all/13/59/28991359.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR