Potassium tetraoxalate dihydrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Oxalic acid, potassium salt dihydratePotassium trihydrogen dioxalate dihydrate

P0475-500G P0475-25G
TCP0475-500GEA 191.87 USD
TCP0475-500G TCP0475-25G
Potassium tetraoxalate dihydrate ≥99.0% (by titrimetric analysis)
Potassium tetraoxalate dihydrate

[for Determination of pH]
CAS Number: 6100-20-5
MDL Number: MFCD00036364
Molecular Formula: C2H2O4
Molecular Weight: 218.16
Purity/Analysis Method: 99.0% (T)
Form: Crystal


Other Product Names / Synonyms: Oxalic Acid Hemipotassium salt, Potassium Tetraoxalate

Formula: C₄H₃KO₈·2H₂O
MW: 254,19 (218,16 anhy)
Storage Temperature: ambient
MDL Number: MFCD00150443
CAS Number: 6100-20-5
EINECS: 204-874-6
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR