Picolinamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-PyridinecarboxamidePyridine-2-carboxamide

P0414-25G
TCP0414-025GEA 107.82 USD
TCP0414-025G
Picolinamide ≥98.0% (by HPLC, titration analysis)
Picolinamide

CAS Number: 1452-77-3
MDL Number: MFCD00023483
Molecular Formula: C6H6N2O
Molecular Weight: 122.13
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 105


Other Product Names / Synonyms: Pyridine-2-carboxamide

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 143 °C
Melting Pt: 105 °C
MDL Number: MFCD00023483
CAS Number: 1452-77-3
https://us.vwr.com/stibo/web/std.lang.all/11/10/28991110.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR