4-Phenyl-1-butyne ≥97.0%

Supplier: TCI America

P0358-5ML
TCP0358-005MLEA 100.49 USD
TCP0358-005ML
4-Phenyl-1-butyne ≥97.0%
4-Phenyl-1-butyne

CAS Number: 16520-62-0
MDL Number: MFCD00041667
Molecular Formula: C10H10
Molecular Weight: 130.19
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 66
Specific Gravity (20/20): 0.93


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Phenethylacetylene, 2-Phenylethylacetylene

Formula: C₁₀H₁₀
MW: 130.19 g/mol
Boiling Pt: 190 °C
Density: 0.93 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00041667
CAS Number: 16520-62-0

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR