Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxyazobenzene ≥98.0%

Supplier: TCI America


P0142-25G
TCP0142-025GEA 220 USD InStock
TCP0142-025G
4-Methoxyazobenzene ≥98.0%
4-Methoxyazobenzene
CAS Number: 2396-60-3
MDL Number: MFCD00039804
Molecular Formula: C13H12N2O
Molecular Weight: 212.25
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 340
Melting point (°C): 56

Other Product Names / Synonyms: p-Phenylazoanisole


Formula: C₁₃H₁₂N₂O
Boiling Pt: 340 °C
Melting Pt: 56 °C
MDL Number: MFCD00039804
CAS Number: 2396-60-3
EINECS: 219-250-9
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty