trans-Cinnamoyl chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (E)-Cinnamoyl chloridetrans-3-Phenylacryloyl Chloride

P0133-25G
TCP0133-25GEA 31.45 USD
TCP0133-25G
trans-Cinnamoyl chloride ≥97.0% (by titrimetric analysis)
trans-Cinnamoyl chloride

CAS Number: 17082-09-6
MDL Number: MFCD00000732
Molecular Formula: C9H7ClO
Molecular Weight: 166.60
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 35


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: trans-3-Phenylacryloyl Chloride

Formula: C₉H₇ClO
MW: 166.61 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00000732
CAS Number: 17082-09-6
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/08/42/28990842.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR