D-(-)-Pantolactone ≥98.0% (by titrimetric analysis)

Synonyms: (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanoneD-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactoneD-(-)-Pantoyl Lactone

P0011-500G
TCP0011-500GEA 478.38 USD
TCP0011-500G
D-(-)-Pantolactone ≥98.0% (by titrimetric analysis)
D-(-)-Pantolactone

CAS Number: 599-04-2
MDL Number: MFCD00005392
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 92
Specific rotation [a]20/D: -50.8 deg (C=2, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, D-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone, D-(-)-Pantoyl Lactone

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00005392
CAS Number: 599-04-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR