Dichloromethyloctylsilane ≥97.0%

Supplier: TCI America

Synonyms: OctylmethyldichlorosilaneDichloro(methyl)(octyl)silane

O0265-25ML
TCO0265-025MLEA 34 USD
TCO0265-025ML
Dichloromethyloctylsilane ≥97.0%
Dichloromethyloctylsilane

CAS Number: 14799-93-0
MDL Number: MFCD00042606
Molecular Formula: C9H20Cl2Si
Molecular Weight: 227.24
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 234
Flash Point (°C): 43
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: n-Octylmethyldichlorosilane

Formula: C₉H₂₀Cl₂Si
MW: 227.25 g/mol
Boiling Pt: 234 °C
Density: 0.97 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00042606
CAS Number: 14799-93-0
UN: 2987
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/05/36/28990536.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR