8-Octanelactam ≥98.0%

Supplier: TCI America

Synonyms: 2-Azacyclononanone8-Octanolactam

O0162-10G
TCO0162-010GEA 148 USD
TCO0162-010G
8-Octanelactam ≥98.0%
8-Octanelactam

CAS Number: 935-30-8
MDL Number: MFCD00003273
Molecular Formula: C8H15NO
Molecular Weight: 141.21
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 151
Melting point (°C): 77


Other Product Names / Synonyms: 2-Azacyclononanone, omega-Caprylolactam, omega-Octanolactam

https://us.vwr.com/stibo/web/std.lang.all/04/54/28990454.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR