1-Naphthol ≥98.0% for biochemical research

Supplier: TCI America

Synonyms: α-Naphtholα-Hydroxynaphthalene1-Hydroxynaphthalene

N0864-5G N0864-1G
TCN0864-5GEA 59.89 USD
TCN0864-5G TCN0864-1G
1-Naphthol ≥98.0% for biochemical research
1-Naphthol

[for Biochemical Research]
CAS Number: 90-15-3
MDL Number: MFCD00003930
Molecular Formula: C10H8O
Molecular Weight: 144.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 97
Flash Point (°C): 125
Lambda max.: 324 nm (MeOH)

Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 147 °C
Melting Pt: 97 °C
Flash Pt: 125 °C
MDL Number: MFCD00003930
CAS Number: 90-15-3
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/00/56/28990056.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR