4-Nitrophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (4-Nitrophenyl)metaboric acid 4-Nitrobenzeneboronic acid Dihydroxy(4-nitrophenyl)borane

N0812-5G N0812-1G
TCN0812-5GEA 111.62 USD
TCN0812-5G TCN0812-1G
4-Nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Nitrophenylboronic acid

CAS Number: 24067-17-2
MDL Number: MFCD00161360
Molecular Formula: C6H6BNO4
Molecular Weight: 166.93
Form: Crystal
Color: White
Melting point (°C): 305


Other Product Names / Synonyms: 4-Nitrobenzeneboronic Acid

Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: 305 °C
MDL Number: MFCD00161360
CAS Number: 24067-17-2
https://us.vwr.com/stibo/web/std.lang.all/00/10/28990010.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR