(4-Nitrobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(4-Nitrobenzyl)triphenylphosphonium bromide
N0701-25G
TCN0701-25GEA
116
USD
TCN0701-25G
(4-Nitrobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(4-Nitrobenzyl)triphenylphosphonium chloride
CAS Number: 2767-70-6
MDL Number: MFCD00032108
Molecular Formula: C25H21BrNO2P
Molecular Weight: 478.33
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 275
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Triphenyl(4-nitrobenzyl)phosphonium Bromide
Formula:
C₂₅H₂₁BrNO₂P MW: 478.33 g/mol Melting Pt: 275 °C |
MDL Number:
MFCD00032108 CAS Number: 2767-70-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...