1-(4-Nitrophenyl)piperazine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0694-5G N0694-25G
TCN0694-5GEA 29.44 USD
TCN0694-5G TCN0694-25G
1-(4-Nitrophenyl)piperazine ≥98.0% (by titrimetric analysis)
1-(4-Nitrophenyl)piperazine

CAS Number: 6269-89-2
MDL Number: MFCD00005961
Molecular Formula: C10H13N3O2
Molecular Weight: 207.23
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 132


Other Product Names / Synonyms: 1-Nitro-4-(1-piperazinyl)benzene

Formula: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00005961
CAS Number: 6269-89-2
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/99/29/28989929.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR