Zinc naphthenate

Synonyms: Naphthenic Acid Zinc Salt

N0341-25G
TCN0341-25GEA 19 USD
TCN0341-25G
Zinc naphthenate
Zinc naphthenate

CAS Number: 12001-85-3
MDL Number: MFCD00147699
Form: Starch Syrup
Color: Colorless
Flash Point (°C): 160


Other Product Names / Synonyms: Naphthenic Acid Zinc Salt

Formula: 2(C₁₁H₇O₂)·Zn
MW: 319.71 g/mol
Flash Pt: 160 °C
MDL Number: MFCD00147699
CAS Number: 12001-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR