TCIAN0338

N0338-500G
TCN0338-500GEA 90.79 USD
TCN0338-500G
TCIAN0338
CAS Number: 61789-51-3
MDL Number: MFCD02094763
Form: Crystal
Color: Purple
Melting point (°C): 140

Other Product Names / Synonyms: Naphthenic Acid Cobalt Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR