5-Nitro-2-furaldehyde diacetate ≥97.0%

Supplier: TCI America

Synonyms: 5-Nitrofurfural Diacetyl Acetal2-(Diacetoxymethyl)-5-nitrofuran5-Nitrofurfurylidene Diacetate(5-Nitrofuran-2-yl)methylene diacetate

N0218-25G
TCN0218-025GEA 145.15 USD
TCN0218-025G
5-Nitro-2-furaldehyde diacetate ≥97.0%
5-Nitro-2-furaldehyde diacetate

CAS Number: 92-55-7
MDL Number: MFCD00003244
Molecular Formula: C9H9NO7
Molecular Weight: 243.17
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 91


Other Product Names / Synonyms: 2-(Diacetoxymethyl)-5-nitrofuran, 5-Nitrofurfural Diacetyl Acetal, 5-Nitrofurfurylidene Diacetate

Formula: C₉H₉NO₇
MW: 243.17 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00003244
CAS Number: 92-55-7
https://us.vwr.com/stibo/web/std.lang.all/93/09/28989309.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR