4-Nitrophenol 0.25% in water

Supplier: TCI America

Synonyms: p-Nitrophenol1-Hydroxy-4-nitrobenzene

N0161-500ML
TCN0161-500MLEA 95.41 USD
TCN0161-500ML
4-Nitrophenol 0.25% in water
4-Nitrophenol

[for pH Determination]
CAS Number: 100-02-7
MDL Number: MFCD00007331
Molecular Weight: 139.11
Form: Clear Liquid
Color: Yellow

Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00007331
CAS Number: 100-02-7
UN: 1663
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/92/58/28989258.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR