β-Naphthol Violet
Supplier: TCI America
Synonyms:
1-(4-Nitrophenylazo)-2-naphthol-3,6-disulfonic Acid Disodium Salt
, 1-(4-Nitrophenylazo)-2-naphthol-3,6-disulphonic Acid Disodium Salt
N0037-1G
TCN0037-001GEA
52.4
USD
TCN0037-001G
β-Naphthol Violet
beta-Naphthol violet
CAS Number: 7143-21-7
MDL Number: MFCD00021457
Molecular Formula: C16H11N3O9S2
Molecular Weight: 497.36
Form: Crystal
Color: Deep Yellow Red
Other Product Names / Synonyms: 1-(4-Nitrophenylazo)-2-naphthol-3,6-disulfonic Acid Disodium Salt
Formula:
C₁₆H₁₁N₃O₉S₂ MW: 497.36 g/mol |
MDL Number:
MFCD00021457 CAS Number: 7143-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...