Methyl-4-iodo-2-methoxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 4-Iodo-2-methoxybenzoic acid methyl esterMethyl 4-iodo-2-methoxybenzoateMethyl-4-iodo-2-methoxybenzoateMethyl 4-iodo-o-anisate5-Iodo-2-(methoxycarbonyl)anisole

M2096-5G M2096-25G
TCM2096-25GEA 164 USD
TCM2096-25G TCM2096-5G
Methyl-4-iodo-2-methoxybenzoate ≥98.0%
Methyl 4-iodo-2-methoxybenzoate

CAS Number: 35387-93-0
MDL Number: MFCD00052925
Molecular Formula: C9H9IO3
Molecular Weight: 292.07
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 95


Other Product Names / Synonyms: 3-Iodo-4-methoxybenzoic Acid Methyl Ester, Methyl 3-Iodo-p-anisate, 3-Iodo-p-anisic Acid Methyl Ester

Formula: C₉H₉IO₃
MW: 292.07 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00052925
CAS Number: 35387-93-0
https://us.vwr.com/stibo/web/std.lang.all/80/53/28988053.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR