Methyl-3-methylcrotonate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3,3-dimethylacrylateMethyl-3,3-dimethylacrylate

M2052-25G
TCM2052-25GEA 111.65 USD
TCM2052-25G
Methyl-3-methylcrotonate ≥98.0%
Methyl 3-methylcrotonate

CAS Number: 924-50-5
MDL Number: MFCD00043940
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 37
Specific Gravity (20/20): 0.94
Lambda max.: 216 nm (CH3CN)


Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: 3,3-Dimethylacrylic Acid Methyl Ester, Methyl 3-Methyl-2-butenoate

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 136 °C
Density: 0.94 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00043940
CAS Number: 924-50-5
UN: 3272
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/80/04/28988004.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR