Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥95.0%

Supplier: TCI America

Synonyms: 2,5-Bis(trifluoromethyl)-3,6-dioxaundecafluorononanoic Acid Methyl Ester

M2030-5G
TCM2030-5GEA 96 USD
TCM2030-5G
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥95.0%
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers)

CAS Number: 26131-32-8
MDL Number: MFCD00054723
Molecular Formula: C10H3F17O4
Molecular Weight: 510.10
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 1.66


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,5-Bis(trifluoromethyl)-3,6-dioxaundecafluorononanoic Acid Methyl Ester

Formula: C₁₀H₃F₁₇O₄
MW: 510.10 g/mol
Boiling Pt: 155 °C
Density: 1.66 (20 °C)
MDL Number: MFCD00054723
CAS Number: 26131-32-8
https://us.vwr.com/stibo/web/std.lang.all/79/78/28987978.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR