3-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-(Methoxycarbonyl)benzeneboronic acid

M1906-5G M1906-25G M1906-1G
TCM1906-5GEA 125.95 USD
TCM1906-5G TCM1906-25G TCM1906-1G
3-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Methoxycarbonyl)phenylboronic acid

CAS Number: 99769-19-4
MDL Number: MFCD02093046
Molecular Formula: C8H9BO4
Molecular Weight: 179.97
Form: Crystal
Color: White
Melting point (°C): 209


Other Product Names / Synonyms: 3-(Methoxycarbonyl)benzeneboronic Acid

Formula: C₈H₉BO₄
Melting Pt: 209 °C
MDL Number: MFCD02093046
CAS Number: 99769-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR