7-Methyl-8-nitroquinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1864-5G
TCM1864-5GEA
327
USD
TCM1864-5G
7-Methyl-8-nitroquinoline ≥98.0% (by GC, titration analysis)
7-Methyl-8-nitroquinoline
CAS Number: 7471-63-8
Molecular Formula: C10H8N2O2
Molecular Weight: 188.19
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 185
Formula:
C₁₀H₈N₂O₂ Melting Pt: 185 °C |
MDL Number:
MFCD00041234 CAS Number: 7471-63-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...